Anti-MAP2K3 Rabbit Monoclonal Antibody [clone: EP555Y]

Lieferant: Abcam
ABCAAB40836-100EA 732 EUR
ABCAAB40836-100
Anti-MAP2K3 Rabbit Monoclonal Antibody [clone: EP555Y]
Antikörper
Type: Primary
Antigen: MAP2K3 (mitogen-activated protein kinase kinase 3)
Clonality: monoclonal
Clone: EP555Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR