Anti-GOLGA3 Goat polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB40837-100EA 634 EUR
ABCAAB40837-100
Anti-GOLGA3 Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: GOLGA3 (golgin A3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR