Anti-EZR Rabbit monoclonal antibody unconjugated [clone: EP886Y]

Lieferant: Abcam
ABCAAB40839-100EA 713 EUR
ABCAAB40839-100
Anti-EZR Rabbit monoclonal antibody unconjugated [clone: EP886Y]
Antikörper
Type: Primary
Antigen: EZR (ezrin)
Clonality: monoclonal
Clone: EP886Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR