Anti-LDB3 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB40840-100EA 666 EUR
ABCAAB40840-100
Anti-LDB3 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: LDB3 (LIM domain binding 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR