Anti-Glutamate receptor 3 Rabbit monoclonal antibody unconjugated [clone: EP813Y]
Lieferant: Abcam
ABCAAB40845-100EA 876 EUR
ABCAAB40845-100
Anti-Glutamate receptor 3 Rabbit monoclonal antibody unconjugated [clone: EP813Y]
Antikörper
Type: Primary
Antigen: Glutamate receptor 3
Clonality: monoclonal
Clone: EP813Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen