Anti-TPH2 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB40846-100EA 696 EUR
ABCAAB40846-100
Anti-TPH2 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: TPH2 (tryptophan hydroxylase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR