Anti-DAZL Goat polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB40848-100EA 672 EUR
ABCAAB40848-100
Anti-DAZL Goat polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: DAZL (deleted in azoospermia-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR