Anti-Asparagine synthetase Rabbit monoclonal antibody unconjugated [clone: EP282Y]

Lieferant: Abcam
ABCAAB40850-100EA 697 EUR
ABCAAB40850-100
Anti-Asparagine synthetase Rabbit monoclonal antibody unconjugated [clone: EP282Y]
Antikörper
Type: Primary
Antigen: Asparagine synthetase
Clonality: monoclonal
Clone: EP282Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR