Anti-VPS37C Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB40851-100EA 657 EUR
ABCAAB40851-100
Anti-VPS37C Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VPS37C (vacuolar protein sorting 37 homolog C (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR