Anti-VPS45 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB40853-100EA 666 EUR
ABCAAB40853-100
Anti-VPS45 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VPS45 (vacuolar protein sorting 45 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR