Anti-SMAD3 Rabbit Monoclonal Antibody [clone: EP568Y]

Lieferant: Abcam
ABCAAB40854-100EA 763 EUR
ABCAAB40854-100
Anti-SMAD3 Rabbit Monoclonal Antibody [clone: EP568Y]
Antikörper
Type: Primary
Antigen: SMAD3 (SMAD family member 3)
Clonality: monoclonal
Clone: EP568Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR