Anti-Macrophage Inflammatory Protein 1 beta Rabbit Monoclonal Antibody [clone: EP494Y]

Lieferant: Abcam
ABCAAB40857-100EA 626 EUR
ABCAAB40857-100
Anti-Macrophage Inflammatory Protein 1 beta Rabbit Monoclonal Antibody [clone: EP494Y]
Antikörper
Type: Primary
Antigen: Macrophage Inflammatory Protein 1 beta
Clonality: monoclonal
Clone: EP494Y
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR