Anti-Elav-type RNA-binding protein ETR3 Rabbit polyclonal antibody unconjugated

ABCAAB50734-100EA 682 EUR
ABCAAB50734-100
Anti-Elav-type RNA-binding protein ETR3 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: Elav-type RNA-binding protein ETR3
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR