Anti-DHX29 Mouse monoclonal antibody unconjugated [clone: 2269C1]
Lieferant: Abcam
ABCAAB50747-100EA 877 EUR
ABCAAB50747-100
Anti-DHX29 Mouse monoclonal antibody unconjugated [clone: 2269C1]
Antikörper
Type: Primary
Antigen: DHX29 (DEAH (Asp-Glu-Ala-His) box polypeptide 29)
Clonality: monoclonal
Clone: 2269C1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human