Anti-EEF2 Mouse monoclonal antibody unconjugated [clone: ELL2A11H2]

Lieferant: Abcam
ABCAAB50748-100EA 882 EUR
ABCAAB50748-100
Anti-EEF2 Mouse monoclonal antibody unconjugated [clone: ELL2A11H2]
Antikörper
Type: Primary
Antigen: EEF2 (eukaryotic translation elongation factor 2)
Clonality: monoclonal
Clone: ELL2A11H2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR