Anti-ALDH1A2 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB75674-100EA 927 EUR
ABCAAB75674-100
Anti-ALDH1A2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ALDH1A2 (aldehyde dehydrogenase 1 family, member A2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse