Anti-ZIP Kinase Rabbit Polyclonal Antibody

Lieferant: Abcam
AB115695-50
ABCAAB115695-50EA 881 EUR
ABCAAB115695-50
Anti-ZIP Kinase Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZIP Kinase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR