Anti-HSD11B2 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB115696-50EA 812 EUR
ABCAAB115696-50
Anti-HSD11B2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HSD11B2 (hydroxysteroid (11-beta) dehydrogenase 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human