Anti-IAPP Mouse Monoclonal Antibody [clone: R10/99]

Lieferant: Abcam
AB115766-50
ABCAAB115766-50EA 848 EUR
ABCAAB115766-50
Anti-IAPP Mouse Monoclonal Antibody [clone: R10/99]
Antikörper
Type: Primary
Antigen: IAPP (islet amyloid polypeptide)
Clonality: monoclonal
Clone: R10/99
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR