Anti-DEFB1 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB115813-100EA 840 EUR
ABCAAB115813-100
Anti-DEFB1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: DEFB1 (defensin, beta 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR