Anti-NOS2 Rabbit Monoclonal Antibody [clone: SP126]

Lieferant: Abcam
AB115819-100
ABCAAB115819-100EA 914 EUR
ABCAAB115819-100
Anti-NOS2 Rabbit Monoclonal Antibody [clone: SP126]
Antikörper
Type: Primary
Antigen: NOS2 (nitric oxide synthase 2, inducible)
Clonality: monoclonal
Clone: SP126
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR