Anti-Digoxin Mouse monoclonal antibody unconjugated [clone: 25P1C9]

Lieferant: Abcam
AB116590-100
ABCAAB116590-100EA 876 EUR
ABCAAB116590-100
Anti-Digoxin Mouse monoclonal antibody unconjugated [clone: 25P1C9]
Antikörper
Type: Primary
Antigen: Digoxin
Clonality: monoclonal
Clone: 25P1C9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR