Anti-RGS1 Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB117077-50EA 859 EUR
ABCAAB117077-50
Anti-RGS1 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RGS1 (regulator of G-protein signaling 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR