Anti-RAD52 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB117097-50EA 840 EUR
ABCAAB117097-50
Anti-RAD52 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: RAD52 (RAD52 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human