Anti-Phosphoserine/threonine Rabbit Polyclonal Antibody

Lieferant: Abcam
AB117253-1000
ABCAAB117253-1000EA 876 EUR
ABCAAB117253-1000
Anti-Phosphoserine/threonine Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Phosphoserine/threonine
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR