Anti-DPAGT1 Rabbit Polyclonal Antibody

Lieferant: Abcam
AB117459-100
ABCAAB117459-100EA 902 EUR
ABCAAB117459-100
Anti-DPAGT1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: DPAGT1 (dolichyl-phosphate (UDP-N-acetylglucosamine) N-acetylglucosaminephosphotransferase 1 (GlcNAc-1-P transferase))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR