Anti-HA tag Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 16B12]

Lieferant: Abcam
ABCAAB117491-100EA 800 EUR
ABCAAB117491-100
Anti-HA tag Mouse Monoclonal Antibody (Peridinin Chlorophyll) [clone: 16B12]
Antikörper
Type: Primary
Antigen: HA tag (HA tag)
Clonality: monoclonal
Clone: 16B12
Conjugation: Peridinin Chlorophyll
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR