Anti-DDDDK tag Mouse Monoclonal Antibody (DyLight® 650) [clone: M2]

Lieferant: Abcam
ABCAAB117492-100EA 840 EUR
ABCAAB117492-100
Anti-DDDDK tag Mouse Monoclonal Antibody (DyLight® 650) [clone: M2]
Antikörper
Type: Primary
Antigen: DDDDK tag (DDDDK tag)
Clonality: monoclonal
Clone: M2
Conjugation: DyLight® 650
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR