Anti-GST Goat Polyclonal Antibody (DyLight® 488)
Lieferant: Abcam
ABCAAB117508-100EA 834 EUR
ABCAAB117508-100
Anti-GST Goat Polyclonal Antibody (DyLight® 488)
Antikörper
Type: Primary
Antigen: GST
Clonality: polyclonal
Clone:
Conjugation: DyLight® 488
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen