Anti-HVCN1 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB117520-50EA 807 EUR
ABCAAB117520-50
Anti-HVCN1 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: HVCN1 (hydrogen voltage-gated channel 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human