Anti-AT11A Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB117577-50EA 763 EUR
ABCAAB117577-50
Anti-AT11A Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: AT11A
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR