Anti-CD59 Mouse Monoclonal Antibody (CF®405M) [clone: VJ1/12.2]

Lieferant: Abcam
ABCAAB117734-50EA 423 EUR
ABCAAB117734-50
Anti-CD59 Mouse Monoclonal Antibody (CF®405M) [clone: VJ1/12.2]
Antikörper
Type: Primary
Antigen: CD59 (CD59 molecule, complement regulatory protein)
Clonality: monoclonal
Clone: VJ1/12.2
Conjugation: CF® 405M Dye
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR