Anti-ZBTB2 Rabbit Polyclonal Antibody
Lieferant: Abcam
ABCAAB117756-20EA 797 EUR
ABCAAB117756-20
Anti-ZBTB2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZBTB2 (zinc finger and BTB domain containing 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse