Anti-PRRC2C Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB117790-100EA 797 EUR
ABCAAB117790-100
Anti-PRRC2C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PRRC2C (proline-rich coiled-coil 2C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR