Anti-EIF4G2 Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB117804-100EA 950 EUR
ABCAAB117804-100
Anti-EIF4G2 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EIF4G2 (eukaryotic translation initiation factor 4 gamma, 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human