Anti-ATL3 Rabbit polyclonal antibody unconjugated

Lieferant: Abcam
ABCAAB117819-100EA 837 EUR
ABCAAB117819-100
Anti-ATL3 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: ATL3 (atlastin GTPase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR