Anti-IDH3G Goat Polyclonal Antibody

Lieferant: Abcam
AB118289-200
ABCAAB118289-200EA 837 EUR
ABCAAB118289-200
Anti-IDH3G Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IDH3G (isocitrate dehydrogenase 3 (NAD+) gamma)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR