Anti-PTPN22 Goat Polyclonal Antibody

Lieferant: Abcam
AB118430-50
ABCAAB118430-50EA 802 EUR
ABCAAB118430-50
Anti-PTPN22 Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PTPN22 (protein tyrosine phosphatase, non-receptor type 22 (lymphoid))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR