Anti-Fibrinogen Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB118488-50EA 853 EUR
ABCAAB118488-50
Anti-Fibrinogen Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Fibrinogen (fibrinogen)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR