Anti-IRGM Rabbit Polyclonal Antibody

Lieferant: Abcam
AB118569-50
ABCAAB118569-50EA 882 EUR
ABCAAB118569-50
Anti-IRGM Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: IRGM (immunity-related GTPase family, M)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR