Anti-ZC3H12C Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB118601-50EA 835 EUR
ABCAAB118601-50
Anti-ZC3H12C Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: ZC3H12C (zinc finger CCCH-type containing 12C)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR