Anti-INO80 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB118787-100EA 842 EUR
ABCAAB118787-100
Anti-INO80 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: INO80 (INO80 complex subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR