Anti-EXOC1 Rabbit polyclonal antibody unconjugated
Lieferant: Abcam
ABCAAB118798-100EA 797 EUR
ABCAAB118798-100
Anti-EXOC1 Rabbit polyclonal antibody unconjugated
Antikörper
Type: Primary
Antigen: EXOC1 (exocyst complex component 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse