Anti-VPS35 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB118838-100EA 938 EUR
ABCAAB118838-100
Anti-VPS35 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: VPS35 (vacuolar protein sorting 35 homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR