Anti-MAOB Goat Polyclonal Antibody

Lieferant: Abcam
ABCAAB118865-50EA 882 EUR
ABCAAB118865-50
Anti-MAOB Goat Polyclonal Antibody
Antikörper
Type: Primary
Antigen: MAOB (monoamine oxidase B)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR