Anti-HGF Mouse Monoclonal Antibody [clone: 7-2]

Lieferant: Abcam
ABCAAB118871-50EA 840 EUR
ABCAAB118871-50
Anti-HGF Mouse Monoclonal Antibody [clone: 7-2]
Antikörper
Type: Primary
Antigen: HGF (hepatocyte growth factor (hepapoietin A; scatter factor))
Clonality: monoclonal
Clone: 42187
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR