Anti-PPP3CC Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB118893-50EA 840 EUR
ABCAAB118893-50
Anti-PPP3CC Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: PPP3CC (protein phosphatase 3, catalytic subunit, gamma isozyme)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR