Anti-Kappa light chain Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: TB28-2]

Lieferant: Abcam
ABCAAB119122-100EA 834 EUR
ABCAAB119122-100
Anti-Kappa light chain Mouse Monoclonal Antibody (APC (Allophycocyanin)) [clone: TB28-2]
Antikörper
Type: Primary
Antigen: Kappa light chain
Clonality: monoclonal
Clone: TB28-2
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR