Anti-SLC2A6 Rabbit Polyclonal Antibody

Lieferant: Abcam
ABCAAB119272-100EA 797 EUR
ABCAAB119272-100
Anti-SLC2A6 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: SLC2A6 (solute carrier family 2 (facilitated glucose transporter), member 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR