Anti-RABL2A Mouse monoclonal antibody [clone: 5A10]

Lieferant: Abcam
ABCAAB119412-100EA 755 EUR
ABCAAB119412-100
Anti-RABL2A Mouse monoclonal antibody [clone: 5A10]
Type: Primary
Antigen: RABL2A (RAB, member of RAS oncogene family-like 2A)
Clonality: monoclonal
Clone: 5A10
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR